  • Search Results
  • Cyclooxygenase 2 and prostaglandin E2 regulate the attachment of calcium oxalate crystals to renal epithelial cells.
Cyclooxygenase 2 and prostaglandin E2 regulate the attachment of calcium oxalate crystals to renal epithelial cells.
International journal of urology : official journal of the Japanese Urological Association (2012-05-30)
Katsuhito Miyazawa, Yoshitaka Takahashi, Nobuyo Morita, Manabu T Moriyama, Takeo Kosaka, Matomo Nishio, Tanihiro Yoshimoto, Koji Suzuki
ABSTRACT
To determine the roles of endogenous cyclooxygenase 2 and prostaglandin E(2) in crystal-cell binding, which is considered to be an important step in the development of intratubular nephrocalcinosis. An expression plasmid for human cyclooxygenase 2 was introduced into Madin-Darby canine kidney cells using the lipofection method. Cyclooxygenase activity was measured using thin-layer chromatography, and the prostaglandin E(2) concentration was determined with an enzyme immunoassay. In addition, crystal attachment was evaluated with a liquid scintillation counter using [(14)C] calcium oxalate monohydrate crystals, and immunohistochemistry and an enzyme immunoassay were used to analyze and quantify the expression of hyaluronan, a crystal-binding molecule. Cyclooxygenase 2-overexpressing Madin-Darby canine kidney cells produced about 10-fold more prostaglandin E(2) than wild-type Madin-Darby canine kidney cells, and their hyaluronan production was also upregulated. The attachment of calcium oxalate monohydrate crystals to cyclooxygenase 2-overexpressing Madin-Darby canine kidney cells was significantly reduced compared with their attachment to wild-type and mock-transfected Madin-Darby canine kidney cells. Pre-incubation of the cyclooxygenase 2-overexpressing cells, as well as the mock-transfected and wild-type cells with the cyclooxygenase 2 selective inhibitor etodolac, increased the cellular attachment of calcium oxalate monohydrate crystals in a dose-dependent manner. These findings suggest that cyclooxygenase 2 expression and the resultant increase in endogenous prostaglandin E(2), leading to increased hyaluronan production, help to prevent nephrocalcinosis by inhibiting the attachment of calcium oxalate monohydrate crystals to the surface of renal epithelial cells.
Sigma-Aldrich
Etodolac
Etodolac for peak identification, European Pharmacopoeia (EP) Reference Standard
Etodolac, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.