  • Search Results
  • Design, synthesis and antiviral potential of 14-aryl/heteroaryl-14H-dibenzo[a,j]xanthenes using an efficient polymer-supported catalyst.
Design, synthesis and antiviral potential of 14-aryl/heteroaryl-14H-dibenzo[a,j]xanthenes using an efficient polymer-supported catalyst.
Molecules (Basel, Switzerland) (2012-06-20)
Kalla Reddi Mohan Naidu, Balam Satheesh Krishna, Mungara Anil Kumar, Palanisamy Arulselvan, Shaik Ibrahim Khalivulla, Ola Lasekan
ABSTRACT
Polyethyleneglycol bound sulfonic acid (PEG-OSO₃H), a chlorosulphonic acid-modified polyethylene glycol was successfully used as an efficient and eco-friendly polymeric catalyst in the synthesis of 14-aryl/heteroaryl-14H-dibenzo[a,j]xanthenes obtained from the reaction of 2-naphthol and carbonyl compounds under solvent-free conditions with short reaction times and excellent yields. The biological properties of these synthesized title compounds revealed that compounds 3b, 3c, 3f and 3i showed highly significant anti-viral activity against tobacco mosaic virus.
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
2-Naphthol, fluorescence indicator, ≥99.0%
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)