MilliporeSigma
  • Peptidergic regulation of expression of genes encoding antioxidant and anti-inflammatory proteins.
Peptidergic regulation of expression of genes encoding antioxidant and anti-inflammatory proteins.
Bulletin of experimental biology and medicine (2012-07-18)
V Kh Khavinson, N S Lin'kova, A V Dudkov, V O Polyakova, I M Kvetnoi
ABSTRACT
Geroprotective peptide T-34 regulates the expression of mRNA for various genes. The development of gastric ulcer is associated with morphological and molecular changes resulting from modulation of the synthesis of antioxidant and anti-inflammatory proteins. Peptide T-34 normalizes the synthesis of these proteins by regulating the expression of the corresponding genes.
Sigma-Aldrich
Cystamine dihydrochloride, BioXtra
Sigma-Aldrich
Cystamine dihydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Cystamine dihydrochloride, 96%