  • Search Results
  • Ethylene bisdithiocarbamate pesticides cause cytotoxicity in transformed and normal human colon cells.
Ethylene bisdithiocarbamate pesticides cause cytotoxicity in transformed and normal human colon cells.
Environmental toxicology and pharmacology (2012-07-25)
Lisa Hoffman, Diane Hardej
ABSTRACT
The effects of the fungicides Maneb, Mancozeb, and Zineb were investigated in transformed colon cells, HT-29, Caco2 and non-transformed cells, CCD-18Co. Significant decreases in viability were observed with Maneb and Mancozeb in HT-29 and CCD-18Co (80-260μM), and Caco2 cells (40-180μM). No significant decreases in viability were observed in all cell types up to 800μM with Zineb. MnCl(2) and ZnCl(2) exposure produced no loss of viability in all cell types up to 400μM. Light microscopy confirmed viability analysis. Lipid peroxidation was observed with Maneb and Mancozeb in cell types tested (60-200μM). Caspase 3/7, 8, and 9 activities were observed with Maneb and Mancozeb in cell types tested (40-200μM). Maneb and Mancozeb treated HT-29 and Caco2 cells demonstrated increases in manganese and zinc concentrations (20-200μM). The lack of toxicity observed with Zineb, MnCl(2), and ZnCl(2) suggests that both the metal moiety and the organic portion of these fungicides together contribute to toxicity.
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.: room temp
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Supelco
Mancozeb, PESTANAL®, analytical standard
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Maneb, PESTANAL®, analytical standard
Sigma-Aldrich
Manganese(II) chloride, flakes, 97%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Supelco
Zineb, PESTANAL®, analytical standard
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis