  • Search Results
  • Lesions of the lateral habenula dissociate the reward-enhancing and locomotor-stimulant effects of amphetamine.
Lesions of the lateral habenula dissociate the reward-enhancing and locomotor-stimulant effects of amphetamine.
Neuropharmacology (2012-07-31)
Anthony J Gifuni, Solmaz Jozaghi, Anne-Catherine Gauthier-Lamer, Sandra M Boye
ABSTRACT
Midbrain dopamine neurons play a key role in goal-directed behaviour as well as in some psychiatric disorders. Recent studies have provided electrophysiological, anatomical and biochemical evidence that the lateral habenula (LHb) exerts strong inhibitory control over midbrain dopamine neurons. However, the behavioural relevance of this inhibitory input is poorly understood. Our aim was to examine the contribution of the LHb to dopamine-sensitive behaviour. Here, we characterized the locomotor-stimulant and reward-enhancing properties of amphetamine in rats with and without neurotoxic lesions of the LHb. Amphetamine-induced forward locomotion and reward were respectively measured in automated activity cages and with intracranial self-stimulation. Adult, male Sprague-Dawley rats were bilaterally infused with ibotenic acid in the LHb and allowed 7-10 days post-operative recovery. The locomotor-stimulant and reward-enhancing properties of amphetamine (0, 0.5 and 1.0 mg/kg, ip) were then tested in different groups of lesioned and sham-lesioned rats. Neurotoxic lesions of the LHb caused a significant enhancement of the locomotor-stimulant effect of amphetamine, an effect not seen following lesions of the medial habenula. Conversely, the reward-enhancing properties of amphetamine did not differ between lesioned and sham-lesioned rats responding for rewarding electrical stimulation of the posterior mesencephalon or the medial forebrain bundle. The dissociation between the locomotor-stimulant and reward-enhancing effects of amphetamine following LHb lesions suggests the contribution of two distinct substrates that are functionally dissociable and differentially sensitive to LHb modulation.
Sigma-Aldrich
Ibotenic acid, ~95%, solid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.