  • Search Results
  • Enantioselective addition of cyclic enol silyl ethers to 2-alkenoyl-pyridine-N-oxides catalysed by Cu(II)-bis(oxazoline) complexes.
Enantioselective addition of cyclic enol silyl ethers to 2-alkenoyl-pyridine-N-oxides catalysed by Cu(II)-bis(oxazoline) complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-08-01)
Alessandro Livieri, Massimo Boiocchi, Giovanni Desimoni, Giuseppe Faita
ABSTRACT
Asymmetric reactions involving (E)-3-aryl-1-(pyridin-2-yl-N-oxide)prop-2-en-1-ones and cyclic enol silyl ethers show good yields and excellent enantioselectivities (up to 99.9 % ee) when catalysed by bis(oxazoline)-Cu(II) complexes. Different reaction pathways can be followed by different enol silyl ethers: with 2-(trimethylsilyloxy)furan, a Mukaiyama-Michael adduct is obtained, whereas a hetero Diels-Alder cycloadduct was formed by using (1,2-dihydronaphthalen-4-yloxy)trimethylsilane. In the latter reaction, the absolute configuration of the product is consistent with a reagent approach to the less hindered Re face of the coordinated substrate in the reactive complex.
Sigma-Aldrich
Pyridine N-oxide, 95%
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.