  • Search Results
  • Renal effects of carprofen and etodolac in euvolemic and volume-depleted dogs.
Renal effects of carprofen and etodolac in euvolemic and volume-depleted dogs.
American journal of veterinary research (2012-08-29)
Kathryn K Surdyk, Dawn L Sloan, Scott A Brown
ABSTRACT
To determine the effects of carprofen and etodolac on renal function in euvolemic dogs and dogs with extracellular fluid volume depletion induced via administration of furosemide. 12 female Beagles. Dogs received a placebo, furosemide, carprofen, etodolac, furosemide and carprofen, and furosemide and etodolac. The order in which dogs received treatments was determined via a randomization procedure. Values of urine specific gravity, various plasma biochemical variables, glomerular filtration rate (GFR [urinary clearance of creatinine]), and renal plasma flow (urinary clearance of para-aminohippuric acid) were determined before and after 8 days of drug administration. A washout time of approximately 12 days was allowed between treatment periods. Administration of furosemide, furosemide and carprofen, and furosemide and etodolac caused changes in urine specific gravity and values of plasma biochemical variables. Administration of carprofen or etodolac alone did not have a significant effect on renal plasma flow or GFR. Concurrent administration of furosemide and carprofen or furosemide and etodolac caused a significant decrease in GFR. After 12-day washout periods, mean values of GFR were similar to values before drug administration for all treatments. Results indicated GFR decreased after 8 days of concurrent administration of furosemide and carprofen or furosemide and etodolac to dogs. Administration of preferential cyclooxygenase-2 inhibitors to dogs with extracellular fluid volume depletion or to dogs treated with diuretics may transiently impair renal function.
Sigma-Aldrich
p-Aminohippuric acid, ≥99%
Sigma-Aldrich
Etodolac
Supelco
Carprofen, VETRANAL®, analytical standard
Etodolac for peak identification, European Pharmacopoeia (EP) Reference Standard
Etodolac, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
p-Aminohippuric acid, ≥98.0% (T)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.