  • Search Results
  • The unravelling of metabolic dysfunctions linked to metal-associated diseases by blue native polyacrylamide gel electrophoresis.
The unravelling of metabolic dysfunctions linked to metal-associated diseases by blue native polyacrylamide gel electrophoresis.
Analytical and bioanalytical chemistry (2012-09-25)
Sungwon Han, Christopher Auger, Zachary Castonguay, Varun P Appanna, Sean C Thomas, Vasu D Appanna
ABSTRACT
Gel electrophoresis is routinely used to separate and analyse macromolecules in biological systems. Although many of these electrophoretic techniques necessitate the denaturing of the analytes prior to their analysis, blue native polyacrylamide gel electrophoresis (BN-PAGE) permits the investigation of proteins/enzymes and their supramolecular structures such as the metabolon in native form. This attribute renders this analytical tool conducive to deciphering the metabolic perturbations invoked by metal toxicity. In this review, we elaborate on how BN-PAGE has led to the discovery of the dysfunctional metabolic pathways associated with disorders such as Alzheimer's disease, Parkinson's disease, and obesity that have been observed as a consequence of exposure to various metal toxicants.
Sigma-Aldrich
Brilliant Blue G solution, Concentrate
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
Brilliant Blue G, 250, for microscopy