  • Search Results
  • Magnetic composites based on metallic nickel and molybdenum carbide: a potential material for pollutants removal.
Magnetic composites based on metallic nickel and molybdenum carbide: a potential material for pollutants removal.
Journal of hazardous materials (2012-10-06)
Raquel V Mambrini, Thales L Fonseca, Anderson Dias, Luiz C A Oliveira, Maria Helena Araujo, Flávia C C Moura
ABSTRACT
New magnetic composites based on metallic nickel and molybdenum carbide, Ni/Mo(2)C, have been produced via catalytic chemical vapor deposition from ethanol. Scanning electron microscopy, thermal analysis, Raman spectroscopy and X-ray diffraction studies suggest that the CVD process occurs in a single step. This process involves the reduction of NiMo oxides at different temperatures (700, 800 and 900°C) with catalytic deposition of carbon from ethanol producing molybdenum carbide on Ni surface. In the absence of molybdenum the formation of Ni/C was observed. The magnetic molybdenum carbide was successfully used as pollutants removal by adsorption of sulfur and nitrogen compounds from liquid fuels and model dyes such as methylene blue and indigo carmine. The dibenzothiofene adsorption process over Ni/Mo(2)C reached approximately 20 mg g(-1), notably higher than other materials described in the literature and also removed almost all methylene blue dye. The great advantage of these carbide composites is that they may be easily recovered magnetically and reused.
Sigma-Aldrich
Dibenzothiophene, 98%
Sigma-Aldrich
Dibenzothiophene, ≥99%
Sigma-Aldrich
Indigo carmine, certified by the Biological Stain Commission, Dye content 85 %
Supelco
Dibenzothiophene, analytical standard
Sigma-Aldrich
Indigo carmine, for microscopy (Bact., Hist.), indicator (pH 11.5-14.0)
Supelco
Indigo carmine, analytical standard