  • Search Results
  • Liquid phase esterification of acetic acid over WO3 promoted β-SiC in a solvent free system.
Liquid phase esterification of acetic acid over WO3 promoted β-SiC in a solvent free system.
Dalton transactions (Cambridge, England : 2003) (2012-10-09)
Gopa Mishra, Gobinda C Behera, S K Singh, K M Parida
ABSTRACT
A series of tungstate promoted β-SiC catalysts was synthesized by a wetness impregnation method. The as synthesized catalysts were unambiguously characterized by XRD, Raman, FTIR, XPS, UV-Vis DRS, TEM, BET surface areas and FE-SEM, and simultaneously the total amount of the acidity of the catalysts was estimated by NH(3)-TPD. The catalytic activities of the synthesized materials were tested in the liquid phase esterification of acetic acid with n-butanol in a solvent free medium. The reaction parameters were optimized to a temperature of 120 °C, molar ratio of butanol and acetic acid of 1:2 and a reaction time of 6 h after performing a number of experiments. Under the optimum conditions, the catalytic esterification revealed a significant effect of 88% conversion with 100% selectivity to butyl acetate in 20 wt% WO(3)/β-SiC. This is the first report on the effective utilization of β-SiC as a catalyst support for liquid phase esterification of acetic acid.
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Supelco
1-Butanol, suitable for HPLC, 99.8%