  • Search Results
  • Catalytic and molecular properties of rabbit liver carboxylesterase acting on 1,8-cineole derivatives.
Catalytic and molecular properties of rabbit liver carboxylesterase acting on 1,8-cineole derivatives.
Natural product communications (2012-10-19)
María H del Loandos, Ana C Muro, Margarita B Villecco, Marcelo F Masman, Paul G M Luiten, Sebastian A Andujar, Fernando D Suvirec, Ricardo D Enriz
ABSTRACT
Rabbit liver carboxylesterase (rCE) was evaluated as the catalyst for the enantioselective hydrolysis of (+/-)-3-endo-acetyloxy-1 ,8-cineole [(+/-)-4], which yields (1S,3S,4R)-(+)-3-acetyloxy-1,8-cineole [(+)-4] and (1R,3R,4S)-(-)-3-hydroxy-1,8-cineole [(-)-3]. Enantioselective asymmetrization of meso-3,5-diacetoxy-1,8-cineol (5) gives (1S,3S,4R,5R)-(-)-3-acetyloxy-5-hydroxy-1,8-cineole (6), with high enantioselectivity. rCE has been chosen to perform both experiments and molecular modeling simulations. Docking simulations combined with molecular dynamics calculations were used to study rCE-catalyzed enantioselective hydrolysis of cineol derivatives. Both compounds were found to bind with their acetyl groups stabilized by hydrogen bond interactions between their oxygen atoms and Ser221.
Sigma-Aldrich
Eucalyptol, 99%
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
1,8-Cineole, primary reference standard
Cineole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Supelco
Eucalyptol, certified reference material, TraceCERT®