MilliporeSigma
  • Temperature-dependent assemblies from a 2-D triple-stranded meso-helical layer to a 3-D chain-layer metal-organic framework.
Temperature-dependent assemblies from a 2-D triple-stranded meso-helical layer to a 3-D chain-layer metal-organic framework.
Dalton transactions (Cambridge, England : 2003) (2012-10-31)
Hong-Guang Jin, Xu-Jia Hong, Jing Li, Yuan-Zhao Yan, Yi-Ting Liu, Shi-Hai Xu, Yue-Peng Cai
ABSTRACT
Two different crystal forms, namely {[Dy2(mpda)3(H2O)2]·H2O}n (1) and [Dy2(mpda)3(H2O)4]n (2), were obtained from the reaction of multidentate 2,6-dimethylpyridine-3,5-dicarboxylic acid (H2mpda) with Dy2O3 at different temperatures. Structural analysis reveals that compound 1 is a 2-D triple-stranded meso-helical layer, and 2 a 3-D chain-layer framework. The fluorescent measure shows that compound 1 is a promising luminescent probe for various metals with a remarkable “on–off” switch of emission, mainly deriving from many pairs of inversely arranged 4-aza bowl-like crown ethers distributed in the 2-D helical plane. Moreover, compound 1 presents noticeable hydrogen-sorption at medium pressure.
Sigma-Aldrich
Dysprosium(III) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium(III) oxide, 99.9% trace metals basis
Sigma-Aldrich
Dysprosium(III) oxide, nanopowder, <100 nm particle size, 99.9% trace metals basis