  • Search Results
  • [The influence of different processing on the content of flavones of Dendranthema morifolium].
[The influence of different processing on the content of flavones of Dendranthema morifolium].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-12-12)
Chuan-shan Jin, De-lin Wu, Nian-jun Yu, Hui-fang Chen, Tong-sheng Wang, Wei Zhang
ABSTRACT
To investigate the effect of different processing methods on the content of luteolin-7-O-beta-D-glucoside, acacetin-7-O-beta-D-glucoside, luteolin, and acacetin of Dendranthema morifolium. The optimal conditions were achieved on phenomenex, synergi Fusion-RP (5 microm, 4.6 mm x 250 mm)analytical column with a gradient mobile phase consisting of methanol and 0.1% phosphoric acid and detection wavelength set at 350 nm. The significant effect of different processing methods on the content of luteolin-7-O-beta-D-glucoside, acacetin-7-O-beta-D-glucoside, luteolin and acacetin was proved. Different processing methods have significant effect on flavonoids of Dendranthema morifolium, it should be paid attention to drying temperature while sulphur fumigation.
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Supelco
Luteolin, analytical standard
Supelco
Luteolin 7-glucoside, analytical standard
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)
Luteolin, primary reference standard
Luteolin 7-glucoside, primary reference standard