  • Search Results
  • Optimization of selenylation conditions for Chinese angelica polysaccharide based on immune-enhancing activity.
Optimization of selenylation conditions for Chinese angelica polysaccharide based on immune-enhancing activity.
Carbohydrate polymers (2012-12-12)
Tao Qin, Jin Chen, Deyun Wang, Yuanliang Hu, Mi Wang, Jing Zhang, The Luong Nguyen, Cui Liu, Xu Liu
ABSTRACT
Chinese angelica polysaccharide (CAP) was extracted by water decoction and ethanol precipitation, purified through eliminating protein by Sevage method and column chromatography of Sephadex G-200, then selenizingly modified by nitric acid-sodium selenite method according to L(9)(3(4)) orthogonal design of three-factors, the usage amount of sodium selenite, reaction temperature and reaction time, at three level to obtain nine selenizing CAPs, sCAP(1)-sCAP(9). Their effects on chicken peripheral lymphocytes proliferation in vitro were compared by MTT assay taking the non-modified CAP as control. The results showed that selenylation modification could significantly enhance the immune-enhancing activity of CAP, sCAP(2) presented best effect and the optimal modification conditions were 200mg of sodium selenite for 500 mg of CAP, the reaction temperature of 70 °C and the reaction time of 6 h.
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium hydrogen selenite, purum p.a., ≥96.0% (RT)