  • Search Results
  • Structural studies of an immunostimulating gluco-arabinan from seeds of Caesalpinia bonduc.
Structural studies of an immunostimulating gluco-arabinan from seeds of Caesalpinia bonduc.
Carbohydrate polymers (2012-12-12)
Eshita Kar Mandal, Soumitra Mandal, Saikat Maity, Birendra Behera, Tapas K Maiti, Syed S Islam
ABSTRACT
A water-soluble gluco-arabinan (PS-II, M(W)∼62 kDa) isolated from the alkaline extract of the endosperm of Caesalpinia bonduc showed the presence of T-Glcp, (1→4)-Glcp, (1→2,3)-Glcp, T-Araf, (1→5)-Araf, (1→2,5)-Araf, and (1→2,3,5)-Araf in a relative proportion of approximately 2:2:2:3:2:1:1. The proposed repeating unit of the polysaccharide possessed a branched backbone of two (1→3)-α-D-glucopyranose followed by four (1→5)-α-L-arabinofuranose residues. In case of two (1→3)-α-D-glucopyranose, branching occurs at O-2 by a same residue terminated by another one at O-4 position. Out of four (1→5)-α-l-arabinofuranose residues, one residue is terminated at O-2 and O-3 by two arabinofuranose residues and another one situated at the adjacent position is terminated at O-2 with same residue, and two (1→5)-α-L-arabinofuranose residues are free from branching and located before and after the two branched arabinofuranose residues. This gluco-arabinan molecule and previously reported arabinan showed similar extent of splenocytes and thymocytes stimulation, but arabinan showed appreciable macrophage activations.
Sigma-Aldrich
DL-Arabinose, ≥98%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.