  • Search Results
  • A new marking technique for peripheral lung nodules avoiding pleural puncture: the intrathoracic stamping method.
A new marking technique for peripheral lung nodules avoiding pleural puncture: the intrathoracic stamping method.
Interactive cardiovascular and thoracic surgery (2012-12-18)
Masaya Kawada, Tetsuyuki Okubo, Saseem Poudel, Yoshinori Suzuki, Yo Kawarada, Shuji Kitashiro, Shunichi Okushiba, Hiroyuki Katoh
ABSTRACT
While performing thoracoscopic wedge resection of the lung, the location of the lesion is generally identified by visual inspection or palpation. When difficulty in identification of the lesion by thoracoscopy is anticipated, preoperative marking is performed. However, complications and technical difficulties plague current marking techniques. To overcome this problem, we designed a new, safe and easy marking technique that avoids pleural puncture, called the intrathoracic stamping method.
Sigma-Aldrich
Indigo carmine, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Indigo carmine, for microscopy (Bact., Hist.), indicator (pH 11.5-14.0)
Supelco
Indigo carmine, analytical standard