  • Search Results
  • [Chemical constituents of ethyl acetate extract from Polygonum perfoliatum].
[Chemical constituents of ethyl acetate extract from Polygonum perfoliatum].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-12-21)
Huan-Bo Cheng, Xin-Qiao Liu, Ke-Li Chen
ABSTRACT
To study the chemical constituents of Polygonum perfoliatum. The chemical constituents of Polygonum perfoliatum were isolated and purified by chromatography on silica gel, Sephadex LH-20 and Semi-preparative HPLC. The structures of compounds were elucidated and identified by physicochemical properties, H-NMR, 13C-NMR and ESI-MS. Ten compounds were isolated and identified as 5-hydroxymethyl-2-furaldEhyde (1), methyl caffeoate (2), protocatechuic aldehyde (3), quercetin (4), pinocernbrin (5), catechin (6), taxifolin (7), taxifolin-3-O-beta-D-xylopyranoside (8), 13-epitorulosal (9), coumarin-7-O-beta-D-glucose glycosidic (10). [corrected] Compounds 6-9 are isolated from this plant for the first time.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Taxifolin, analytical standard
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Taxifolin, primary reference standard
Supelco
Ethyl acetate, HPLC grade, 99.8%