  • Search Results
  • Evaluation of the potential for diacetylenes as reporter molecules in 3D micelle gel dosimetry.
Evaluation of the potential for diacetylenes as reporter molecules in 3D micelle gel dosimetry.
Physics in medicine and biology (2013-01-17)
A T Nasr, T Olding, L J Schreiner, K B McAuley
ABSTRACT
Radiochromic micelle gel dosimeters are promising for three-dimensional (3D) radiation dosimetry because they can be read out by optical CT techniques and they have superior spatial stability compared to polymer and Fricke gel dosimeters. This study evaluates the use of diacetylenes as reporter molecules in micelle gel dosimeters. Several gels containing pentacosa-10,12-diynoic acid (PCDA) emulsified using sodium dodecyl sulfate (SDS) changed from colourless to blue upon irradiation. Unfortunately, all phantoms that experienced a colour change were turbid and would be unsuitable for 3D dosimetry. Two techniques (use of organic solvent and aqueous-phase additives) were successful in increasing colloidal stability to prevent the turbidity problem, but none of the resulting transparent gels changed colour in response to radiation. Transparent PCDA emulsions were prepared using NaOH solutions with no SDS or other emulsifier, but these transparent emulsions also did not change colour. Only turbid gels and emulsions with precipitated particles responded to radiation. These results indicate that the colour change was due to the oligomerization within precipitated PCDA crystals, and that liquid-phase emulsified PCDA did not undergo oligomerization. As a result, PCDA is not suitable for use in micelle gel dosimeters, and other radiochromic reporter molecules will need to be identified.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M