  • Search Results
  • Adiponectin enhances calcium dependency of mouse bladder contraction mediated by protein kinase Cα expression.
Adiponectin enhances calcium dependency of mouse bladder contraction mediated by protein kinase Cα expression.
The Journal of pharmacology and experimental therapeutics (2013-02-01)
Koji Nobe, Akiko Fujii, Kiyomi Saito, Takaharu Negoro, Yoshio Ogawa, Yasuko Nakano, Terumasa Hashimoto, Kazuo Honda
ABSTRACT
Adiponectin is an adipose tissue-secreted protein and is a multifunctional adipocytokine. However, the association of adiponectin with bladder contraction has not been investigated. In this study, the adiponectin-sense transgenic mouse (Adip-Sen mouse; age, 16-24 weeks; male) and age-matched controls (C57Bl mouse) were studied. The Adip-Sen mouse showed a significant increase in plasma adiponectin levels (56.2%; P < 0.01), compared with those in the C57Bl mouse, without affecting other lipid parameters. Isometric force development in bladder smooth muscle tissues were detected using an organ-bath system. Although carbachol (CCh)-induced (0.1-100 µM) time- and dose-dependent contractions in Adip-Sen mouse bladder were slightly enhanced, compared with those in the C57Bl mouse during a low range (0.3-1.0 µM) of CCh, differences could not be detected with other CCh concentrations. However, the reduction in contraction under Ca(2+)-replaced conditions was significantly different between Adip-Sen and C57Bl mice (94.1 and 66.3% of normal contraction, respectively; n = 5). A parameter of Ca(2+) sensitivity, the relation between intracellular Ca(2+) concentration and contraction, was increased in the Adip-Sen mouse, compared with that in the C57B1 mouse. This Ca(2+) dependency in the Adip-Sen mouse was reduced by a protein kinase C (PKC) inhibitor, but not by a Rho kinase inhibitor. Expression of the calcium-dependent isoform of PKC, PKCα, was increased in the Adip-Sen mouse bladder, and CCh-induced phosphorylation of PKCα was also enhanced, compared with those in the C57Bl mouse. In conclusion, adiponectin is associated with bladder smooth muscle contraction, which involves an increase in Ca(2+) dependency of contraction mediated by PKCα expression.
Product Description
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Carbachol, European Pharmacopoeia (EP) Reference Standard