  • Search Results
  • Struvite pyrolysate recycling combined with dry pyrolysis for ammonium removal from wastewater.
Struvite pyrolysate recycling combined with dry pyrolysis for ammonium removal from wastewater.
Bioresource technology (2013-02-12)
Rongtai Yu, Jinju Geng, Hongqiang Ren, Yanru Wang, Ke Xu
ABSTRACT
The dry pyrolysis of magnesium ammonium phosphate (MAP) with NaOH powder for ammonium release was investigated, as well as the utility of MAP pyrolysate recycling. The identities of the MAP pyrolysate and its derivatives were experimentally validated. The results showed that the pyrolysate was amorphous magnesium hydrogen phosphate (MgHPO4) and magnesium pyrophosphate (Mg2P2O7). The best molar ratio of sodium hydroxide (NaOH) powder to ammonium was 1:1, at 110°C for 3h. The optimum pH for pyrolysate recycling was 9.5. The ammonia removal ratio could be maintained above 80% with MAP pyrolysate recycling. Seed crystal inoculation increased the rate of MAP crystallization by 20.86%, as well as the MAP grain size (2.08nm with seeding versus 1.72nm without). MAP particle size with NaOH treatment decreased: d(0.5)=19.34μm versus d(0.5)=30.35μm for direct pyrolysis. The results demonstrated that crystal growth was controlled by adding NaOH during MAP pyrolysis.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh