  • Search Results
  • Effects of NH4Cl and MgCl(2) on pretreatment and xylan hydrolysis of miscanthus straw.
Effects of NH4Cl and MgCl(2) on pretreatment and xylan hydrolysis of miscanthus straw.
Carbohydrate polymers (2013-02-13)
Kyeong Eop Kang, Don-Hee Park, Gwi-Taek Jeong
ABSTRACT
This study investigated the effects of NH(4)Cl and MgCl(2) on pretreatment and xylan hydrolysis of miscanthus straw for biofuels production. It was observed that increasing the pretreatment temperature decreased the remaining solid, increased the enzymatic digestibility, and increased the xylan removal. When 0.2-5.0% NH(4)Cl and MgCl(2) were employed in pretreatments, increasing the inorganic salt concentration slightly diminished the remaining solid, though the enzymatic digestibility was enhanced. Under the higher-than-2% condition, no xylan remained in the solid residues after pretreatment. With pretreatment time, the remaining solid slightly decreased, but the enzymatic digestibility was increased. Moreover, xylan removal was linearly increased to 15 min, after which it was completely hydrolyzed. Overall, these results indicated that pretreatment by 2% NH(4)Cl or MgCl(2) at 185 °C for 15 min completely hydrolyzes the xylan of miscanthus straw. In scanning electron microscopy (SEM) images, the physical surface of the miscanthus straw showed an apparently damaged surface area and exposure of the internal structure after pretreatment with NH(4)Cl and MgCl(2) by SEM.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M