MilliporeSigma
  • Polyion-counterion interactions in sodium carboxymethylcellulose-ethylene glycol-water ternary solutions.
Polyion-counterion interactions in sodium carboxymethylcellulose-ethylene glycol-water ternary solutions.
Carbohydrate polymers (2013-02-13)
Ramesh Sharma, Chanchal Das, Sanjay Dahal, Bijan Das
ABSTRACT
Polyion-counterion interactions in sodium carboxymethylcellulose-ethylene glycol-water ternary system have been investigated with special reference to their variations with the polyelectrolyte concentration, the medium relative permittivity and the temperature using electrical conductance measurements. The experimental data have been analyzed on the basis of a model for semidilute polyelectrolyte conductivity which takes into account the scaling arguments proposed by Dobrynin et al. Concentration-dependent moderate counterion condensation (24-33%) was observed. Counterion condensation is found to be spontaneous which depends upon the EG-content of the medium and the temperature. A major proportion of the current is transported by the carboxymethylcellulose polyions. The results further demonstrated that the monomer units experience more frictional resistance in solution as the EG-content increases or as the temperature decreases.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~700,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~90,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol 5 M solution