  • Search Results
  • Application of ionic liquid-based microwave-assisted extraction of malachite green and crystal violet from water samples.
Application of ionic liquid-based microwave-assisted extraction of malachite green and crystal violet from water samples.
Journal of separation science (2013-02-26)
Zhanqi Gao, Tingfeng Liu, Xiaoju Yan, Cheng Sun, Huan He, Shaogui Yang
ABSTRACT
A simple, environment friendly and efficient technique, ionic liquid-based microwave-assisted extraction was first used to determine malachite green and crystal violet (CV) from water samples coupled to HPLC. The key parameters influencing extraction efficiency were investigated, such as the type of ionic liquids, the volume of ionic liquid, extraction time, and so on. Under the optimum conditions, good reproducibility of the extraction performance was obtained (RSD, 1.0% for malachite green (MG) and 5.9% for CV, n = 5). Good linearity (0.10-25 μg L(-1)) was observed with correlation coefficients between 0.9991 and 0.9964. The detection limits of MG and CV were 0.080 and 0.030 μg L(-1), respectively. The proposed method had been successfully applied to determine MG and CV in real water samples with recoveries ranging from 95.4 to 102.8%. Compared with the previous technologies, the proposed method required less extraction time (2 min), and provided lower detection limits and higher enrichment factors. Moreover, there were no volatile and hazardous organic solvents released. Based on these simple, environment friendly, rapid, and highly efficient results, the proposed approach provides a new and promising alternative for simultaneously extracting trace amounts of MG and CV from water.
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal violet solution
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal violet solution
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
Crystal violet solution, 1%, aqueous solution
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Sigma-Aldrich
Gentian Violet solution, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard