  • Search Results
  • Preparation of hybrid soda-lime/quartz glass chips with wettability-patterned channels for manipulation of flow profiles in droplet-based analytical systems.
Preparation of hybrid soda-lime/quartz glass chips with wettability-patterned channels for manipulation of flow profiles in droplet-based analytical systems.
Analytica chimica acta (2013-03-05)
Zeqing Bai, Qiaohong He, Shanshi Huang, Xianqiao Hu, Hengwu Chen
ABSTRACT
Profile switching of two-phase flows is often required in microfluidic systems. Manipulation of flow profiles can be realized by control of local surface energy of micro channel through wettability-patterning of channel surface. This article presents a facile approach for wettability-patterning of the micro channels of glass chips. Commercially available octadecyltrichlorosilane (OTS) was used to hydrophobilize the channels via the formation of OTS self-assembly monolayer (SAM), and a UV-source that mainly emits deep UV-light of 254 and 185 nm was employed to degrade the in-channel formed OTS-SAM. The architecture of soda-lime glass/quartz glass hybrid chip was designed to facilitate the deep UV-light effective degrading the OTS-SAM. The established approach, together with the side-by-side laminar-flow patterning technique, was applied to prepare various finely patterned channel networks for different tasks of flow profile switching. The micro device capable of conducting the profile switch from W/O droplets to two separated continuous phases was demonstrated to perform on-chip quick liquid-liquid extraction for the determination of partition coefficients of pharmaceuticals.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Soda lime, ACS reagent, granular, +100 mesh particle size
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Silicon dioxide, ~99%, 0.5-10 μm (approx. 80% between 1-5 μm)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Soda lime, p.a., with indicator, pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC