  • Search Results
  • Removal of carbamazepine from aqueous solution by adsorption on fly ash-amended soil.
Removal of carbamazepine from aqueous solution by adsorption on fly ash-amended soil.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-03-20)
Maria K Swarcewicz, Justyna Sobczak, Waldemar Paździoch
ABSTRACT
Carbamazepine (CBZ), nonbiodegradable pharmaceutical residue, has become an emerging pollutant in several aquatic environments. The effectiveness of the mixture of soil and fly ash (FA) in adsorbing CBZ from aqueous solution has been studied as well as agitation time, FA content, initial CBZ concentration and desorption as a function of FA content. The adsorption kinetics fits a hyperbolic or pseudo-second-order model. The maximum adsorbed amounts for natural soil and a mixture of soil/FA ranged from 77 to 158 mg kg(-1). Rate constants were considered relatively low (4.15-15.59 × 10(-4) kg mg(-1) min). The logarithmic form of the Freundlich equation gave a linearity and the Kf constants increased with the increase of FA content in adsorbent mixtures and with the affinity between the adsorbent surface and adsorbed solute. The mean removed amounts of CBZ by adsorption batch experiments in a soil mixture with 30% FA content were up to 92.8% for coal FA and 33% in natural soil. This work proved that the mixture of the coal FA and soil can be used as an efficient adsorbent material for removal of CBZ from water.
Product Description
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder