  • Search Results
  • High resolution spectroscopy of the Ar-D2O and Ar-HDO molecular complexes in the near-infrared range.
High resolution spectroscopy of the Ar-D2O and Ar-HDO molecular complexes in the near-infrared range.
The Journal of chemical physics (2013-03-22)
K Didriche, T Földes
ABSTRACT
Three rovibrational bands of Ar-D2O and two rovibrational bands of Ar-HDO were observed in the 1.5 μm range by continuous wave cavity ringdown spectroscopy. Their analyses led to the determination of rotational constants for the upper states and vibrational shifts indicating that the potential energy surface is only slightly affected by the vibrational excitation. Some Coriolis couplings were identified. The observed lines were fitted to retrieve a 3.5 ns lifetime of the upper state, showing that even with a triple or double excitation quanta in the water subunit, the Ar-D2O and Ar-HDO complexes are long-lived species.
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Argon, ≥99.998%