MilliporeSigma
GdBr3: CE in a glass wafer as a nuclear radiation monitor.
Health physics (2013-03-28)
Zhitao Kang, Robert Rosson, M Brooke Barta, Jason Nadler, Brent Wagner, Bernd Kahn
ABSTRACT
A glass wafer that contains cerium-activated gadolinium-based scintillator has been tested as a nuclear radiation monitor. The detector is prepared by mixing powdered gadolinium and cerium (3+) bromides with alumina, silica, and lithium fluoride, melting the mixture at 1,400°C, and then quenching and annealing the glass. The resulting clear glass matrix emits stimulated blue light that can be collected by a conventional photomultiplier tube. Spectral analysis of radionuclides with this detector shows the energy peaks for alpha particles, the energy continuum for beta particles, the Compton continuum and full-energy peaks for gamma rays, and an energy continuum with specific reaction-product peaks for neutrons. Energy resolution for the 5.5-MeV alpha particle and 0.662-MeV gamma-ray peaks is about 20%. This resolution, although threefold poorer than for single-crystal NaI(Tl) scintillators, contributes to radionuclide identification and quantification. Application of this detector to radiation monitoring is proposed, as well as approaches for improving light collection and energy resolution that will facilitate radionuclide identification and monitoring, especially for alpha particles, beta particles, and low-energy gamma rays.
Sigma-Aldrich
Lithium fluoride, powder, -300 mesh
Sigma-Aldrich
Lithium fluoride, powder, <100 μm, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Lithium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Lithium fluoride, Precipitated, 99.995%
Sigma-Aldrich
Lithium-6Li fluoride, 95 atom % 6Li, 99% (CP)
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O