  • Search Results
  • Catharanthine dilates small mesenteric arteries and decreases heart rate and cardiac contractility by inhibition of voltage-operated calcium channels on vascular smooth muscle cells and cardiomyocytes.
Catharanthine dilates small mesenteric arteries and decreases heart rate and cardiac contractility by inhibition of voltage-operated calcium channels on vascular smooth muscle cells and cardiomyocytes.
The Journal of pharmacology and experimental therapeutics (2013-03-28)
Ashok Jadhav, Wenbin Liang, Peter C Papageorgiou, Ahmed Shoker, Selliah C Kanthan, John Balsevich, Andrew S Levy, Scott Heximer, Peter H Backx, Venkat Gopalakrishnan
ABSTRACT
Catharanthine is a constituent of anticancer vinca alkaloids. Its cardiovascular effects have not been investigated. This study compares the in vivo hemodynamic as well as in vitro effects of catharanthine on isolated blood vessels, vascular smooth muscle cells (VSMCs), and cardiomyocytes. Intravenous administration of catharanthine (0.5-20 mg/kg) to anesthetized rats induced rapid, dose-dependent decreases in blood pressure (BP), heart rate (HR), left ventricular blood pressure, cardiac contractility (dP/dt(max)), and the slope of the end-systolic pressure-volume relationship (ESPVR) curve. Catharanthine evoked concentration-dependent decreases (I(max) >98%) in endothelium-independent tonic responses of aortic rings to phenylephrine (PE) and KCl (IC(50) = 28 µM for PE and IC(50) = 34 µM for KCl) and of third-order branches of the small mesenteric artery (MA) (IC(50) = 3 µM for PE and IC(50) = 6 µM for KCl). Catharanthine also increased the inner vessel wall diameter (IC(50) = 10 µM) and reduced intracellular free Ca(2+) levels (IC(50) = 16 µM) in PE-constricted MAs. Patch-clamp studies demonstrated that catharanthine inhibited voltage-operated L-type Ca(2+) channel (VOCC) currents in cardiomyocytes and VSMCs (IC(50) = 220 µM and IC(50) = 8 µM, respectively) of MA. Catharanthine lowers BP, HR, left ventricular systolic blood pressure, and dP/dt(max) and ESPVR likely via inhibition of VOCCs in both VSMCs and cardiomyocytes. Since smaller vessels such as the third-order branches of MAs are more sensitive to VOCC blockade than conduit vessels (aorta), the primary site of action of catharanthine for lowering mean arterial pressure appears to be the resistance vasculature, whereas blockade of cardiac VOCCs may contribute to the reduction in HR and cardiac contractility seen with this agent.
Product Description
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis