  • Search Results
  • Boundary of the nucleotide-binding domain of Streptococcus ComA based on functional and structural analysis.
Boundary of the nucleotide-binding domain of Streptococcus ComA based on functional and structural analysis.
Biochemistry (2013-03-29)
Seiji Ishii, Takato Yano, Akihiro Okamoto, Takeshi Murakawa, Hideyuki Hayashi
ABSTRACT
The ATP-binding cassette (ABC) transporter ComA is a key molecule essential for the first step of the quorum-sensing system of Streptococcus. The nucleotide binding domains (NBD) of Streptococcus mutans ComA with different N termini, NBD1 (amino acid residues 495-760), NBD2 (517-760), and NBD3 (528-760), were expressed, purified, and characterized. The shortest NBD3 corresponds to the region commonly defined as NBD in the database searches of ABC transporters. A kinetic analysis showed that the extra N-terminal region conferred a significantly higher ATP hydrolytic activity on the NBD at a neutral pH. Gel-filtration, X-ray crystallography, and mutational analyses suggest that at least four to five residues beyond the N-terminal boundary of NBD3 indeed participate in stabilizing the protein scaffold of the domain structure, thereby facilitating the ATP-dependent dimerization of NBD which is a prerequisite to the catalysis. These findings, together with the presence of a highly conserved glycine residue in this region, support the redefinition of the N-terminal boundary of the NBD of these types of ABC exporters.
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Alanine, ≥99.0% (NT)
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C