  • Search Results
  • FIP200 is required for maintenance and differentiation of postnatal neural stem cells.
FIP200 is required for maintenance and differentiation of postnatal neural stem cells.
Nature neuroscience (2013-04-02)
Chenran Wang, Chun-Chi Liang, Z Christine Bian, Yuan Zhu, Jun-Lin Guan
ABSTRACT
Despite recent studies showing that inhibition of autophagy depletes the hematopoietic stem cell pool and increases intracellular reactive oxygen species (ROS), it remains unknown whether autophagy is essential in the maintenance of other stem cells. Moreover, it is unclear whether and how the aberrant ROS increase causes depletion of stem cells. Here we report that ablation of FIP200 (also known as Rb1cc1), a gene essential for autophagy induction in mammalian cells, results in a progressive loss of neural stem cells (NSCs) and impairment in neuronal differentiation specifically in the postnatal brain, but not the embryonic brain, in mice. The defect in maintaining the postnatal NSC pool was caused by p53-dependent apoptotic responses and cell cycle arrest. However, the impaired neuronal differentiation was rescued by treatment with the antioxidant N-acetylcysteine but not by p53 inactivation. These data reveal that FIP200-mediated autophagy contributes to the maintenance and functions of NSCs through regulation of oxidative state.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%