MilliporeSigma
Asymmetric synthesis of (-)-martinellic acid.
Organic letters (2013-04-06)
Stephen G Davies, Ai M Fletcher, James A Lee, Thomas J A Lorkin, Paul M Roberts, James E Thomson
ABSTRACT
A high-yielding total asymmetric synthesis of (-)-martinellic acid is reported. The conjugate addition of lithium (R)-N-allyl-N-(α-methyl-4-methoxybenzyl)amide to tert-butyl (E)-3-[2'-(N,N-diallylamino)-5'-bromophenyl]propenoate and alkylation of the resultant β-amino ester have been used as the key steps to install the C(9b) and C(3a) stereogenic centers, respectively, and a highly diastereoselective Wittig reaction/intramolecular Michael addition was then used to create the C(4) stereogenic center within this tricyclic molecular architecture.
Product Description
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, granular, 99% trace metals basis