  • Search Results
  • Exposure to cocaine regulates inhibitory synaptic transmission in the nucleus accumbens.
Exposure to cocaine regulates inhibitory synaptic transmission in the nucleus accumbens.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-04-19)
Mami Otaka, Masago Ishikawa, Brian R Lee, Lei Liu, Peter A Neumann, Ranji Cui, Yanhua H Huang, Oliver M Schlüter, Yan Dong
ABSTRACT
Medium spiny neurons (MSNs) within the nucleus accumbens shell (NAc) function to gate and prioritize emotional/motivational arousals for behavioral output. The neuronal output of NAc MSNs is mainly determined by the integration of membrane excitability and excitatory/inhibitory synaptic inputs. Whereas cocaine-induced alterations at excitatory synapses and membrane excitability have been extensively examined, the overall functional output of NAc MSNs following cocaine exposure is still poorly defined because little is known about whether inhibitory synaptic input to these neurons is affected by cocaine. Here, our results demonstrate multidimensional alterations at inhibitory synapses in NAc neurons following cocaine self-administration in rats. Specifically, the amplitude of miniature IPSCs (mIPSCs) was decreased after 21 d withdrawal from 5 d cocaine self-administration. Upon re-exposure to cocaine after 21 d withdrawal, whereas the amplitude of mIPSCs remained downregulated, the frequency became significantly higher. Furthermore, the reversal potential of IPSCs, which was not significantly altered during withdrawal, became more hyperpolarized upon cocaine re-exposure. Moreover, the relative weight of excitatory and inhibitory inputs to NAc MSNs was significantly decreased after 1 d cocaine withdrawal, increased after 21 d withdrawal, and returned to the basal level upon cocaine re-exposure after 21 d withdrawal. These results, together with previous results showing cocaine-induced adaptations at excitatory synapses and intrinsic membrane excitability of NAc MSNs, may provide a relatively thorough picture of the functional state of NAc MSNs following cocaine exposure.
Product Description
SAFC
L-Valine
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
NBQX hydrate, powder, ≥98% (HPLC)
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
Cocaine hydrochloride
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Supelco
L-Valine, certified reference material, TraceCERT®
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
NBQX disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Cocaine free base