  • Search Results
  • Effect of methanol extract of Bombax malabaricum leaves on nitric oxide production during inflammation.
Effect of methanol extract of Bombax malabaricum leaves on nitric oxide production during inflammation.
Acta poloniae pharmaceutica (2013-04-26)
Emdad Hossain, Debjani Sarkar, Mitali Chatterjee, Saikat Chakraborty, Subhash C Mandal, Jayanta K Gupta
ABSTRACT
In different parts of India, Bombax malabaricum DC. (Family: Bombacaceae), a lofty deciduous tree with large leaves, is traditionally used in inflammation. The aim of the present study was to confirm its antiinflammatory activity and to search for the possible mechanism of action for methanol extract of Bombax malabaricum leaves (MEBM). The anti-inflammatory activity of MEBM was evaluated in a carrageenan-induced model of acute inflammation. As inflammation usually involves increased nitric oxide (NO) production, effect of MEBM on lipopolysaccharide-induced NO production in mouse peritoneal macrophages was studied to evaluate its possible mechanism of action. It was found that MEBM was non-toxic up to a dose of 2 g/kg for mice and rats, orally. MEBM (100, 200, and 400 mg/kg) significantly reduced carrageenan-induced rat paw edema (p < 0.05, p < 0.01, p < 0.001, respectively). In mice peritoneal macrophages, the IC50 for MEBM was 258.33 +/- 6.96 microg/mL and it was non-toxic up to 125 microg/mL. MEBM (0-100 microg/mL) reduced lipopolysaccharide-induced NO production in macrophages in a dose-dependent fashion (p < 0.001). Hence, MEBM possesses antiinflammatory activity, mediated through inhibition of NO production.
Sigma-Aldrich
Carrageenan, suitable for gel preparation
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%