MilliporeSigma
Stabilization of source-separated human urine by chemical oxidation.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-05-10)
Yang Zhang, Zifu Li, Yuan Zhao, Shuangling Chen, Ibrahim Babatunde Mahmood
ABSTRACT
The inhibitory effect of ozone and hydrogen peroxide (HP) on urea hydrolysis in stored urine was investigated and compared. Ozone showed less effect on urea hydrolysis due to the complicated composition of urine (including a large amount of urease-producing bacteria) and bacteria regeneration. Ozone concentration and total heterotrophic bacteria analysis demonstrated that residual ozone concentration decreased by 43% within 15 hr from 13.50 to 7.72 mg/L in the one-time ozonation urine test, and finally completely decomposed within 4 days. In addition, bacteria regenerated quickly after ozone completely decomposed. However, HP showed a significant effect on inhibiting urea hydrolysis not only in stored urine but also in fecal-contaminated urine. The suitable doses of applied HP to inhibit urea hydrolysis in stored urine, concentrations of 0.5 and 1.0 g feces per liter of fecal-contaminated urine, were 0.03, 0.16 and 0.23 mol/L, respectively. The urea concentrations after 2 months stored were 7,145, 7,109 and 7,234 mg/L, respectively.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Urea, European Pharmacopoeia (EP) Reference Standard