  • Search Results
  • The use of elemental sulfur as an alternative feedstock for polymeric materials.
The use of elemental sulfur as an alternative feedstock for polymeric materials.
Nature chemistry (2013-05-23)
Woo Jin Chung, Jared J Griebel, Eui Tae Kim, Hyunsik Yoon, Adam G Simmonds, Hyun Jun Ji, Philip T Dirlam, Richard S Glass, Jeong Jae Wie, Ngoc A Nguyen, Brett W Guralnick, Jungjin Park, Arpád Somogyi, Patrick Theato, Michael E Mackay, Yung-Eun Sung, Kookheon Char, Jeffrey Pyun
ABSTRACT
An excess of elemental sulfur is generated annually from hydrodesulfurization in petroleum refining processes; however, it has a limited number of uses, of which one example is the production of sulfuric acid. Despite this excess, the development of synthetic and processing methods to convert elemental sulfur into useful chemical substances has not been investigated widely. Here we report a facile method (termed 'inverse vulcanization') to prepare chemically stable and processable polymeric materials through the direct copolymerization of elemental sulfur with vinylic monomers. This methodology enabled the modification of sulfur into processable copolymer forms with tunable thermomechanical properties, which leads to well-defined sulfur-rich micropatterned films created by imprint lithography. We also demonstrate that these copolymers exhibit comparable electrochemical properties to elemental sulfur and could serve as the active material in Li-S batteries, exhibiting high specific capacity (823 mA h g(-1) at 100 cycles) and enhanced capacity retention.
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis