  • Search Results
  • Cobalamin deficiency manifested with seizures, mood oscillations, psychotic features and reversible dementia in the absence of typical neurologic and hematologic signs and symptoms: a case report.
Cobalamin deficiency manifested with seizures, mood oscillations, psychotic features and reversible dementia in the absence of typical neurologic and hematologic signs and symptoms: a case report.
Collegium antropologicum (2013-05-24)
Maja Vilibić, Vlado Jukić, Andelko Vidović, Petrana Brecić
ABSTRACT
Cobalamin deficiency is associated with a wide spectrum of hematologic, neurologic, gastroenterologic and psychiatric disorders or symptoms. We report a case of a 50-year-old man with complex partial seizures with secondary generalization, mood oscillations and psychotic symptoms alternating with confusion and reversible dementia secondary to cobalamin deficiency in the absence of typical neurologic and/or hematologic symptoms and signs. Exclusion of epilepsy, acute, atrophic or expansive lesion of central nervous system and usual etiology associated with reversible dementia (infectious diseases, an endocrine etiology and deficiency of vitamins other than cobalamin); finding of cobalamin deficiency only and complete neuropsychiatric recovery after substitution, confirmed etiology. Typical and atypical psychiatric manifestations due to cobalamin deficiency that precede neurologic and/or hematologic signs and symptoms can recover completely after adequate replacement therapy.
Sigma-Aldrich
Diazepam
Supelco
Diazepam solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Haloperidol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Carbamazepine, powder