  • Search Results
  • Dentin hypersensitivity after teeth bleaching with in-office systems. Randomized clinical trial.
Dentin hypersensitivity after teeth bleaching with in-office systems. Randomized clinical trial.
American journal of dentistry (2013-06-04)
Javier Martin, Eduardo Fernandez, Valeria Bahamondes, Andrea Werner, Klaus Elphick, Osmir Batista Oliveira, Gustavo Moncada
ABSTRACT
To comparatively and prospectively compare in a randomized clinical trial, dentin hypersensitivity after treatment with three in-office bleaching systems, based on hydrogen peroxide at different concentrations, with and without light source activation. 88 individuals were included according to inclusion and exclusion criteria. Subjects were randomly divided into the following three treatment groups: Group 1 was treated with three 15-minute applications of hydrogen peroxide at 15% with titanium dioxide (Lase Peroxide Lite) that was light-activated (Light Plus Whitening Lase) with five cycles of 1 minute and 30 seconds each cycle, giving a total treatment time of 45 minutes; Group 2 was treated with three 10-minute applications of hydrogen peroxide at 35% (Lase Peroxide Sensy), activated by light (LPWL) same activation cycles than Group 1, with a total treatment time of 30 minutes; Group 3 was treated with only one application for 45 minutes of hydrogen peroxide at 35% (Whitegold Office) without light activation. Each subject underwent one session of bleaching on the anterior teeth according to the manufacturers' instructions. Dentin sensitivity was recorded with a visual analogue scale (VAS) at baseline, immediately after, and at 7 and 30 days after treatment using a stimulus of an evaporative blowing triple syringe for 3 seconds on the upper central incisors from a distance of 1 cm. A Kruskal-Wallis test followed by Mann-Whitney test was performed for statistical analysis. All groups showed increased sensitivity immediately after treatment. Group 1 displayed less changes relative to baseline with no significant differences (P = 0.104). At 7 and 30 days after treatment, a comparison of VAS values indicated no significant differences between all groups (P = 0.598 and 0.489, respectively).
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer