  • Search Results
  • [Reseach of oxidative stress induces aging in rabbit intervertebral disc nucleus pulposus cells injured by H2O2].
[Reseach of oxidative stress induces aging in rabbit intervertebral disc nucleus pulposus cells injured by H2O2].
Zhongguo gu shang = China journal of orthopaedics and traumatology (2013-07-13)
Jian Xie, Pei-Jian Tong, Le-Tian Shan, Cheng-liang Wu
ABSTRACT
To study the effect of H2O2 on the morphological pattern,vitality,proliferation,cycle period of rabbit intervertebral disc nucleus pulposus cells. Ten New Zealand white rabbits (2 to 3 kg, female) were used for isolating nucleus pulposus cells under sterilized condition. The culture solution with 15% FBS and DMEM/F12 (1:1) was applied for cell cultivation. After 90% cell fusion, the first generation was obtain and stimulated by H2O2 with different concentrations of 0 micromol/L (control group), 130 micromol/L,216 p.mol/L,360 Ipmol/L, 600 micromol/L,and 1000 micromol/L. Compared with the control group, there was little difference of the biological property (P>0.05) in 130 micromol/L and 216 micromol/L H202-treated groups. When the concentration of H2O2 attained 360 micromol/L, 600 micromol/L, and 1 000 micromol/L, the cells suffered aging,with increased cell vacuoles,decreased proliferation,and aging:related increase of 13-galactosidase dyeing. The cell cycle of many nucleus pulposus cells was blocked in G1 stage other than entering S stage. With increasing H2O2 concentrations, the aging degree was increased. A certain concentration of H202 could induce early aging of nucleus pulposus cells,resulting in biological abnormalities of these cells.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis