  • Search Results
  • Electrical and optical properties of indium zinc oxide (IZO) thin films by continuous composition spread.
Electrical and optical properties of indium zinc oxide (IZO) thin films by continuous composition spread.
Journal of nanoscience and nanotechnology (2013-07-19)
J J Lee, J S Kim, S J Yoon, Y S Cho, J W Choi
ABSTRACT
Indium zinc oxide (IZO) films were deposited on glass substrate at room temperature using off-axis RF sputtering-continuous composition spread (CCS) system. The full range composition of IZO films were controlled by the deposition rate and thickness profiles of In2O3 and ZnO target. The structural, electrical and optical properties of IZO thin films were measured as functions of position. IZO thin film had the lowest resistivity and highest carrier concentration at the position of 15 mm (5.02 x 10(-4) omega cm, 3.9 x 10(20)/cm3). And IZO thin film had high transmittance in visible region at measured all positions. This study has investigated to explore the new composition of IZO films using CCS system.
Sigma-Aldrich
Indium(III) oxide, 99.998% trace metals basis
Sigma-Aldrich
Indium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Indium(III) oxide, nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, 40 wt. % in ethanol, <130 nm particle size
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −60 mesh, 99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis