  • Search Results
  • Morphological modification of alpha-MnO2 catalyst for use in Li/air batteries.
Morphological modification of alpha-MnO2 catalyst for use in Li/air batteries.
Journal of nanoscience and nanotechnology (2013-07-19)
Min-Sik Park, Jae-Hun Kim, Ki Jae Kim, Goojin Jeong, Young-Jun Kim
ABSTRACT
Single crystal alpha-MnO2 nanowires and nanopowders have been successfully synthesized in order to facilitate a comparison of their catalytic activity for use in Li-air batteries. The importance of the morphological modification of the alpha-MnO2 catalyst for facilitating electrochemical reactions between Li and O2 is addressed. Distinctive catalytic activity of alpha-MnO2 is observed, which is in line with its different morphologies. The catalytic activity significantly affects the reversible capacity of Li-air batteries. A high aspect ratio, large surface area and good dispersibility of alpha-MnO2 in the nanowire form are advantageous providing larger active surfaces for promoting the fundamental reactions in Li-air batteries. We also introduce a robustly designed air-electrode composed of highly porous carbon and nanostructured alpha-MnO2 catalysts, with employs a metal foam current collector to ensure sufficient air-permeability and to maximize electronic conduction during cycles. Our suggestions should prove helpful in forming a basis for further investigations in developing advanced Li-air batteries.
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%
Sigma-Aldrich
Manganese(IV) oxide, ReagentPlus®, ≥99%