  • Search Results
  • Preparation and characterization of gold-decorated graphite nanosheet composites.
Preparation and characterization of gold-decorated graphite nanosheet composites.
Journal of nanoscience and nanotechnology (2013-07-19)
Jungsoo Kim, Dae Geun Nam, Weon Tae Oh
ABSTRACT
Some composites of gold nanoparticles and graphite nanosheets were prepared by electrostatic interaction, and structurally and electrochemically characterized using X-ray diffraction, X-ray photoelectron spectroscopy, UVNis spectroscopy, transmission electron microscopy, and cyclic-voltammetry. Pristine graphite was chemically treated using aqueous acid solution, and dispersed inpoly(diallyldimethylammonium) chloride aqueous solution to prepare positively charged graphite nanosheets. The gold nanoparticles (GNPs) in this work were stabilized by sodium dodecyl sulfate, poly(sodium 4-styrene sulfonate), or poly(vinylpyrrolidone). Gold nanoparticles and graphite nanosheet composites with gold nanoparticles showed the characteristic surface plasmon band at -530 nm. The electrochemical properties of the graphite nanosheet composites with gold nanoparticles were studied by cyclic voltammetry, in which reduction potential and reduction current of gold nanoparticles were strongly dependent on the gold-wrapped stabilizer in the composites.
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold coated microscope slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated microscope slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis