  • Search Results
  • Visible light-induced photocatalytic activity of Bi2O3 prepared via microwave-assisted method.
Visible light-induced photocatalytic activity of Bi2O3 prepared via microwave-assisted method.
Journal of nanoscience and nanotechnology (2013-08-02)
Xinjuan Liu, Likun Pan, Jinliang Li, Kai Yu, Zhuo Sun
ABSTRACT
Bi2O3 was successfully synthesized via microwave-assisted reaction of Bi2O3 precursor in aqueous solution using a microwave system. Their morphologies, structures and photocatalytic performances in the degradation of methylene blue (MB) were characterized by scanning electron microscopy, transmission electron microscopy, energy dispersive X-ray spectroscopy, X-ray diffraction spectroscopy, photoluminescence spectroscopy and UV-vis absorption spectroscopy, respectively. The results show that Bi2O3 synthesized at pH value of 7 exhibits an optimal photocatalytic performance with the MB degradation rate of 76% at 240 min under visible light irradiation due to its higher visible light absorption and comparatively low electron-hole pair recombination.
Sigma-Aldrich
Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, purum, ≥98.0% (KT)
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis