  • Search Results
  • In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts.
In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts.
Acta poloniae pharmaceutica (2013-08-08)
Mohammed Auwal Ibrahim, Neil Anthony Koorbanally, Md Shahidul Islam
ABSTRACT
The present study was conducted to investigate the anti-oxidative activities of different solvent extracts of Cassia singueana parts. Our results indicate that all the extracts have reducing power (Fe3+ --> Fe2+) and DPPH radical scavenging abilities. However, the ethyl acetate extract of the stem bark has the highest total reducing power whilst the ethanol extract of the stem bark has more potent free radical scavenging activity than all the other extracts. The ethyl acetate extract of the stem bark exhibited more powerful hydroxyl radical scavenging activity than other extracts whilst the aqueous extract of the leaves displayed more potent nitric oxide inhibition activity than other extracts. The GC-MS analysis of the ethyl acetate extract of the stem bark and the ethanol extract of the root and leaves indicated that several aromatic compounds, including phenolics, fatty acids, amino acids and triterpenoids were present in these extracts. Data from this study suggest that the parts of C. singueana possessed anti-oxidative activities and can be used as a potential alternative medicine for oxidative stress related non-communicable chronic diseases. Further experimental and clinical studies in this regard are warranted.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology