  • Search Results
  • Identification of compounds in mouse urine vapor by gas chromatography and mass spectrometry.
Identification of compounds in mouse urine vapor by gas chromatography and mass spectrometry.
Mechanisms of ageing and development (1980-06-01)
K Miyashita, A B Robinson
ABSTRACT
Eighty compounds in normal mouse urine vapor are identified, 61 of which have not previously been reported in normal mouse urine. Two of the ten most strongly age-correlated substances in the mouse urine vapor are confirmed as 3-hydroxy-3-methylbutene and trans-3-hepten-2-one.
Sigma-Aldrich
2-Methyl-3-buten-2-ol, 98%
Sigma-Aldrich
Coconut ketone solution, 10 wt. % in ethanol, natural (US)