  • Search Results
  • Antagonist pharmacology of adenosine A2B receptors from rat, guinea pig and dog.
Antagonist pharmacology of adenosine A2B receptors from rat, guinea pig and dog.
European journal of pharmacology (2003-09-05)
John R Fozard, Francois Baur, Cedric Wolber
ABSTRACT
We have sought evidence for species differences between adenosine A2B receptors by comparing the potencies of eight adenosine receptor antagonists, representing four different chemical classes, at the native adenosine A2B receptors which mediate relaxation of smooth muscle from rat colon, guinea pig aorta and dog saphenous vein. In all three assays, the antagonists caused parallel rightward shifts in the concentration-response curves to NECA and there was no depression of the maximum responses. There were highly significant correlations between the pKB values on each of the three receptors. However, the pKB values of 8-SPT (8-p-(sulphophenyl)theophylline), XAC (8-[-[[[[(2-aminoethyl)amino]-carbonyl]methyl]oxy]phenyl]-1,3-dipropylxanthine), CGS 15943 (9-chloro-2,2-(furanyl)[1,2,4]triazolo[1,5-c]quinazolin-5-amine) and CGH 2473 N-[4-(3,4-dichloro-phenyl)-5-pyridin-4-yl-thiazol-2-yl]-acetamide) for the dog receptor exceeded by at least 0.5 log units the pKB values at the rat and guinea pig sites. Our data indicate species differences between the rat and guinea pig adenosine A2B receptors on the one hand and the dog adenosine A2B receptor on the other with respect to antagonist pharmacology.
Sigma-Aldrich
CGS-15943, solid