MilliporeSigma
  • Downregulation of cell-free miR-198 as a diagnostic biomarker for lung adenocarcinoma-associated malignant pleural effusion.
Downregulation of cell-free miR-198 as a diagnostic biomarker for lung adenocarcinoma-associated malignant pleural effusion.
International journal of cancer (2013-01-29)
Hye-Suk Han, Jieun Yun, Sung-nam Lim, Joung-Ho Han, Ki Hyeong Lee, Seung Taik Kim, Min-Ho Kang, Seung-Myoung Son, Yong-Moon Lee, Song-Yi Choi, Seok Joong Yun, Wun-Jae Kim, Ok-Jun Lee
ABSTRACT
Circulating cell-free microRNAs (miRNAs) are potential cancer biomarkers. The aim of this study was to identify miRNAs that are differentially expressed between benign pleural effusion (BPE) and lung adenocarcinoma-associated malignant pleural effusion (LA-MPE). The expression level of cell-free miRNA was investigated in 107 patients with pleural effusion. Microarrays were used to screen 160 miRNAs in a discovery set comprising 20 effusion samples (ten BPEs and ten LA-MPEs). Real-time quantitative reverse transcription polymerase chain reaction (qRT-PCR) was used to validate the profiling results obtained for the discovery set and those obtained for a validation set comprising 42 BPEs and 45 LA-MPEs. The area under the receiver operating characteristic curve (AUC) was used to evaluate the diagnostic performance of the identified miRNAs and other common tumor markers, such as carcinoembryonic antigen (CEA) and cytokeratin fragment (CYFRA) 21-1. Microarray profiling showed that miR-198 was significantly downregulated in LA-MPE compared with BPE (p = 0.002). The miRNA microarray analysis results were confirmed by qRT-PCR (p < 0.001) using the validation set. The AUCs for miR-198, CEA and CYFRA 21-1 in the validation set were 0.887, 0.898 and 0.836, respectively. The diagnostic performance of miR-198 was comparable with that of CEA, but better than that of CYFRA 21-1. The AUC for all three markers combined was 0.926 (95% confidence interval, 0.843-0.973) with a sensitivity of 89.2% and a specificity of 85.0%. The present study suggests that cell-free miR-198 from patients with pleural effusion might have diagnostic potential for differentiating LA-MPE from BPE.