  • Search Results
  • Restoration of spermatogenesis in dibromochloropropane (DBCP)-treated rats by hormone suppression.
Restoration of spermatogenesis in dibromochloropropane (DBCP)-treated rats by hormone suppression.
Toxicological sciences : an official journal of the Society of Toxicology (2003-09-30)
Marvin L Meistrich, Gene Wilson, Karen L Porter, Ilpo Huhtaniemi, Gunapala Shetty, Gladis A Shuttlesworth
ABSTRACT
The exposure of men to the nematocide dibromochloropropane (DBCP) has caused prolonged oligo- and azoospermia, which occasionally reverses spontaneously. We recently demonstrated that in testes of rats treated with a dose of DBCP sufficient to reduce the percentage of tubules producing differentiating germ cells (tubule differentiation index, TDI) to 20%, the tubules lacking differentiating cells contained type A spermatogonia. To determine whether these type A spermatogonia could be stimulated to differentiate, as had been demonstrated previously in other models of toxicant-induced sterility, we suppressed intratesticular testosterone and serum follicle stimulating hormone (FSH) levels with the GnRH agonist Lupron (leuprolide). When the GnRH agonist was given for 10 weeks starting immediately after DBCP exposure, the TDI was maintained at 94%. Even when GnRH-agonist treatment was stopped at week 10, the TDI remained between 65 and 80% 10 weeks later. Late spermatid counts averaged 10 x 10(6) per testis for the GnRH-agonist-treated rats at week 20 compared with 1.7 x 10(6) per testis in rats treated with only DBCP. To determine whether spermatogonial differentiation could be stimulated after the TDI had declined to below 30%, we initiated GnRH-agonist treatment 6 weeks after DBCP exposure. The GnRH treatment increased the TDI to 53% at week 16. These results indicate that, if the same principles apply to humans, suppression of testosterone may be applied to restore spermatogenesis in men rendered azoospermic by DBCP or other reproductive toxicants.
Product Description
Supelco
1,2-Dibromo-3-chloropropane solution, certified reference material, 200 μg/mL in methanol
Supelco
1,2-Dibromo-3-chloropropane, PESTANAL®, analytical standard
Sigma-Aldrich
1,2-Dibromo-3-chloropropane, 97%
Supelco
1,2-Dibromo-3-chloropropane solution, certified reference material, 5000 μg/mL in methanol
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.