  • Search Results
  • Synthesis of nucleoside libraries on solid support. II. 2,6,8-Trisubstituted purine nucleosides using 8-bromoguanosine as precursor.
Synthesis of nucleoside libraries on solid support. II. 2,6,8-Trisubstituted purine nucleosides using 8-bromoguanosine as precursor.
Nucleosides, nucleotides & nucleic acids (2004-03-27)
Yung-hyo Koh, Michael B Landesman, Roberto Amador, Frank Rong, Haoyun An, Zhi Hong, Jean-Luc Girardet
ABSTRACT
A series of 2,6,8-trisubstituted purine nucleoside libraries was prepared by parallel solid-phase synthesis using 8-bromoguanosine as a common synthetic precursor. Polystyrene-methoxytrityl chloride resin was linked to the N2 or O5' position of the guanosine analogues. 8-Bromoguanosine was derivatized at the C8 position via carbon-carbon bond formation. Nucleophilic aromatic substitution at C2 and/or C6 positions with various amines produced two series of purine nucleoside libraries with very diverse substitution.