  • Search Results
  • Functional characterization and expression analysis of the androgen receptor in zebrafish (Danio rerio) testis.
Functional characterization and expression analysis of the androgen receptor in zebrafish (Danio rerio) testis.
Reproduction (Cambridge, England) (2008-05-13)
P P de Waal, D S Wang, W A Nijenhuis, R W Schulz, J Bogerd
ABSTRACT
The biological activity of androgens, important for male sexual differentiation and development, is mediated by the androgen receptor (AR) that binds to specific DNA recognition sites regulating the transcription of androgen target genes. We investigated androgen production by adult zebrafish testis tissue, and identified 11beta-hydroxyandrostenedione, 11-ketoandrostenedione (OA), and 11-ketotestosterone (11-KT) as main products, and hence potential ligands, for the zebrafish Ar. These androgens were then included in the pharmacological characterization of the zebrafish Ar. The zebrafish Ar responded well in terms of binding and transactivation to synthetic androgens as well as to testosterone and 11-KT, and reasonably well to OA and androstenedione. In situ hybridization analysis of zebrafish testis revealed that ar mRNA expression was detected in the subpopulation of Sertoli cells contacting early spermatogonia.
Sigma-Aldrich
Adrenosterone, 98%