  • Search Results
  • Kynuramines, metabolites of melatonin and other indoles: the resurrection of an almost forgotten class of biogenic amines.
Kynuramines, metabolites of melatonin and other indoles: the resurrection of an almost forgotten class of biogenic amines.
Journal of pineal research (2009-07-04)
Rüdiger Hardeland, Dun-Xian Tan, Russel J Reiter
ABSTRACT
Kynuramines represent their own class of biogenic amines. They are formed either by decarboxylation of kynurenines or pyrrole ring cleavage of indoleamines. N(2)-formylated compounds formed in this last reaction can be deformylated either enzymatically by arylamine formamidases or hemoperoxidases, or photochemically. The earlier literature mainly focussed on cardiovascular effects of kynuramine, 5-hydroxykynuramine and their N(1),N(1)-dimethylated analogs, including indirect effects via release of catecholamines or acetylcholine and interference with serotonin receptors. After the discovery of N(1)-acetyl-N(2)-formyl-5-methoxykynuramine (AFMK) and N(1)-acetyl-5-methoxykynuramine (AMK) as major brain metabolites of melatonin, these compounds became of particular interest. They were shown to be produced enzymatically, pseudoenzymatically, by various free radical-mediated and via photochemical processes. In recent years, AFMK and AMK were shown to scavenge reactive oxygen and nitrogen species, thereby forming several newly discovered 3-indolinone, cinnolinone and quinazoline compounds, and to protect tissues from damage by reactive intermediates in various models. AMK is of special interest due to its properties as a potent cyclooxygenase inhibitor, NO scavenger forming a stable nitrosation product, inhibitor and/or downregulator of neuronal and inducible NO synthases, and a mitochondrial metabolism modulator. AMK easily interacts with aromates, forms adducts with tyrosyl and tryptophanyl residues, and may modify proteins.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.